Assay Details
Gene Symbol: SMIM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005563216.2 | XP_005563273.1 | 2 - 3 | 228 | 78 | |
XM_005563217.2 | XP_005563274.1 | 2 - 3 | 260 | 78 | ||
XM_005563218.2 | XP_005563275.1 | 2 - 3 | 492 | 78 | ||
XM_005563219.2 | XP_005563276.1 | 2 - 3 | 194 | 78 | ||
XM_005563220.2 | XP_005563277.1 | 2 - 3 | 602 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
