Assay Details
Gene Symbol: HSCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005567706.2 | XP_005567763.1 | 1 - 2 | 316 | 77 | |
XM_015457340.1 | XP_015312826.1 | 1 - 2 | 317 | 77 | ||
XM_015457341.1 | XP_015312827.1 | 1 - 2 | 319 | 77 | ||
XM_015457342.1 | XP_015312828.1 | 1 - 2 | 316 | 77 | ||
XM_015457343.1 | XP_015312829.1 | 1 - 2 | 316 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
