Assay Details
Gene Symbol: APOBEC3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015457181.1 | XP_015312667.1 | 4 - 5 | 896 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
