Assay Details
Gene Symbol: LITAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005591265.2 | XP_005591322.1 | 2 - 3 | 336 | 78 | |
XM_005591266.2 | XP_005591323.1 | 2 - 3 | 245 | 78 | ||
XM_005591267.2 | XP_005591324.1 | 2 - 3 | 358 | 78 | ||
XM_005591269.2 | XP_005591326.1 | 2 - 3 | 447 | 78 | ||
XM_005591272.2 | XP_005591329.1 | 2 - 3 | 401 | 78 | ||
XM_015442607.1 | XP_015298093.1 | 2 - 3 | 245 | 78 | ||
XM_015442608.1 | XP_015298094.1 | 2 - 3 | 243 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
