Assay Details
Gene Symbol: CDC42SE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005541942.2 | XP_005541999.1 | 4 - 5 | 685 | 101 | |
XM_005541943.2 | XP_005542000.1 | 5 - 6 | 848 | 101 | ||
XM_005541944.2 | XP_005542001.1 | 5 - 6 | 748 | 101 | ||
GenBank mRNA | AB174710.1 | - | 3 - 4 | 457 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
