Assay Details
Gene Symbol: ISCU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283497.1 | NP_001270426.1 | 2 - 3 | 254 | 67 | |
GenBank mRNA | AB174249.1 | - | 2 - 3 | 254 | 67 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
