Assay Details
Gene Symbol: MUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283081.1 | NP_001270010.1 | 1 - 2 | 260 | 136 | |
XM_015451020.1 | XP_015306506.1 | 1 - 2 | 584 | 136 | ||
GenBank mRNA | AB168850.1 | - | 1 - 2 | 260 | 136 |

More Information
Set Membership: |
![]() ![]() |
