Assay Details
Gene Symbol: MUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283081.1 | NP_001270010.1 | 2 - 3 | 348 | 80 | |
XM_015451020.1 | XP_015306506.1 | 2 - 3 | 672 | 80 | ||
XM_015451024.1 | XP_015306510.1 | 2 - 3 | 960 | 80 | ||
GenBank mRNA | AB168850.1 | - | 2 - 3 | 348 | 80 |
