Assay Details
Gene Symbol: MPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283575.1 | NP_001270504.1 | 4 - 5 | 395 | 96 | |
GenBank mRNA | AB172577.1 | - | 4 - 5 | 395 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
