Assay Details
Gene Symbol: EIF3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284779.1 | NP_001271708.1 | 3 - 4 | 541 | 109 | |
GenBank mRNA | AB169626.1 | - | 3 - 4 | 541 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
