Assay Details
Gene Symbol: R3HCC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283748.1 | NP_001270677.1 | 6 - 7 | 2136 | 80 | |
GenBank mRNA | AB169220.1 | - | 6 - 7 | 2136 | 80 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: R3HCC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283748.1 | NP_001270677.1 | 6 - 7 | 2136 | 80 | |
GenBank mRNA | AB169220.1 | - | 6 - 7 | 2136 | 80 |