Assay Details
Gene Symbol: PERP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285001.1 | NP_001271930.1 | 2 - 3 | 431 | 93 | |
GenBank mRNA | AB170355.2 | - | 2 - 3 | 431 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
