Assay Details
Gene Symbol: UBE2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283170.1 | NP_001270099.1 | 4 - 5 | 410 | 93 | |
XM_005545648.2 | XP_005545705.1 | 4 - 5 | 706 | 93 | ||
GenBank mRNA | AB178995.1 | - | 4 - 5 | 410 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
