Assay Details
Gene Symbol: RELT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283176.1 | NP_001270105.1 | 2 - 3 | 183 | 74 | |
XM_005579035.2 | XP_005579092.1 | 2 - 3 | 959 | 74 | ||
XM_005579037.2 | XP_005579094.1 | 2 - 3 | 324 | 74 | ||
XM_005579038.2 | XP_005579095.1 | 2 - 3 | 959 | 74 | ||
XM_005579039.2 | XP_005579096.1 | 2 - 3 | 959 | 74 | ||
GenBank mRNA | AB046039.1 | - | 2 - 3 | 183 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
