Assay Details
Gene Symbol: SH3BGRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283717.1 | NP_001270646.1 | 2 - 3 | 322 | 82 | |
XM_015443725.1 | XP_015299211.1 | 2 - 3 | 674 | 82 | ||
GenBank mRNA | AB168749.1 | - | 2 - 3 | 322 | 82 |
