Assay Details
Gene Symbol: KDELR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285233.1 | NP_001272162.1 | 4 - 5 | 716 | 93 | |
GenBank mRNA | AB171648.1 | - | 4 - 5 | 716 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
