Assay Details
Gene Symbol: MRPL49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283495.1 | NP_001270424.1 | 1 - 2 | 110 | 73 | |
GenBank mRNA | AB169536.1 | - | 1 - 2 | 124 | 73 | |
AB170521.1 | - | 1 - 2 | 110 | 73 | ||
AB174650.1 | - | 1 - 2 | 110 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
