Assay Details
Gene Symbol: MRPL49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283495.1 | NP_001270424.1 | 2 - 3 | 252 | 106 | |
GenBank mRNA | AB169536.1 | - | 2 - 3 | 266 | 106 | |
AB170521.1 | - | 2 - 3 | 252 | 106 | ||
AB174650.1 | - | 2 - 3 | 252 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
