Assay Details
Gene Symbol: PPIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284819.1 | NP_001271748.1 | 3 - 4 | 466 | 111 | |
XM_015448794.1 | XP_015304280.1 | 2 - 3 | 462 | 111 | ||
GenBank mRNA | AB168546.1 | - | 3 - 4 | 466 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
