Assay Details
Gene Symbol: CDK2AP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285183.1 | NP_001272112.1 | 1 - 2 | 136 | 74 | |
GenBank mRNA | AB174635.1 | - | 1 - 2 | 136 | 74 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
