Assay Details
Gene Symbol: CDK2AP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285183.1 | NP_001272112.1 | 2 - 3 | 225 | 78 | |
XM_005572548.2 | XP_005572605.1 | 2 - 3 | 247 | 78 | ||
XM_005572549.2 | XP_005572606.1 | 2 - 3 | 201 | 78 | ||
GenBank mRNA | AB174635.1 | - | 2 - 3 | 225 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
