Assay Details
Gene Symbol: B3GAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283657.1 | NP_001270586.1 | 2 - 3 | 205 | 104 | |
XM_015434112.1 | XP_015289598.1 | 1 - 2 | 875 | 104 | ||
XM_015434114.1 | XP_015289600.1 | 1 - 2 | 117 | 104 | ||
GenBank mRNA | AB170993.1 | - | 2 - 3 | 205 | 104 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
