Assay Details
Gene Symbol: B3GAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283657.1 | NP_001270586.1 | 4 - 5 | 741 | 87 | |
XM_015434111.1 | XP_015289597.1 | 4 - 5 | 1482 | 87 | ||
XM_015434112.1 | XP_015289598.1 | 3 - 4 | 1411 | 87 | ||
XM_015434113.1 | XP_015289599.1 | 4 - 5 | 1650 | 87 | ||
XM_015434114.1 | XP_015289600.1 | 3 - 4 | 653 | 87 | ||
GenBank mRNA | AB170993.1 | - | 4 - 5 | 741 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
