Assay Details
Gene Symbol: POLE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284168.1 | NP_001271097.1 | 1 - 2 | 246 | 69 | |
XM_015433428.1 | XP_015288914.1 | 1 - 2 | 267 | 69 | ||
GenBank mRNA | AB174423.1 | - | 1 - 2 | 246 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
