Assay Details
Gene Symbol: YARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283100.1 | NP_001270029.1 | 1 - 2 | 788 | 101 | |
GenBank mRNA | AB173582.1 | - | 1 - 2 | 788 | 101 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
