Assay Details
Gene Symbol: NDUFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283245.1 | NP_001270174.1 | 2 - 3 | 267 | 98 | |
GenBank mRNA | AB171884.1 | - | 2 - 3 | 267 | 98 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
