Assay Details
Gene Symbol: RND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283504.1 | NP_001270433.1 | 2 - 3 | 561 | 84 | |
GenBank mRNA | AB170323.1 | - | 2 - 3 | 561 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
