Assay Details
Gene Symbol: AQP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283536.1 | NP_001270465.1 | 1 - 2 | 442 | 86 | |
XM_015447295.1 | XP_015302781.1 | 1 - 2 | 733 | 86 | ||
GenBank mRNA | AB170709.1 | - | 1 - 2 | 442 | 86 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
