Assay Details
Gene Symbol: STMN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283705.1 | NP_001270634.1 | 2 - 3 | 190 | 59 | |
XM_015457625.1 | XP_015313111.1 | 2 - 3 | 359 | 59 | ||
GenBank mRNA | AB169859.1 | - | 2 - 3 | 192 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
