Assay Details
Gene Symbol: PHYHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283782.1 | NP_001270711.1 | 3 - 4 | 702 | 80 | |
XM_005562778.2 | XP_005562835.1 | 3 - 4 | 909 | 80 | ||
GenBank mRNA | AB174614.1 | - | 3 - 4 | 702 | 80 |
