Assay Details
Gene Symbol: MEA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283914.1 | NP_001270843.1 | 2 - 3 | 298 | 99 | |
XM_005552961.2 | XP_005553018.1 | 2 - 3 | 418 | 99 | ||
XM_005552962.2 | XP_005553019.1 | 2 - 3 | 399 | 99 | ||
XM_015448770.1 | XP_015304256.1 | 2 - 3 | 735 | 99 | ||
GenBank mRNA | AB168204.1 | - | 2 - 3 | 298 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
