Assay Details
Gene Symbol: NDUFA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283352.1 | NP_001270281.1 | 3 - 4 | 398 | 86 | |
GenBank mRNA | AB171873.1 | - | 3 - 4 | 398 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
