Assay Details
Gene Symbol: AP5M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283327.1 | NP_001270256.1 | 2 - 3 | 994 | 78 | |
XM_015453248.1 | XP_015308734.1 | 2 - 3 | 817 | 78 | ||
XM_015453249.1 | XP_015308735.1 | 2 - 3 | 1028 | 78 | ||
XM_015453250.1 | XP_015308736.1 | 2 - 3 | 812 | 78 | ||
XM_015453251.1 | XP_015308737.1 | 2 - 3 | 1133 | 78 | ||
GenBank mRNA | AB169123.1 | - | 2 - 3 | 994 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
