Assay Details
Gene Symbol: SQOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284042.1 | NP_001270971.1 | 4 - 5 | 534 | 95 | |
XM_005559446.2 | XP_005559503.1 | 4 - 5 | 899 | 95 | ||
XM_005559447.2 | XP_005559504.1 | 4 - 5 | 610 | 95 | ||
GenBank mRNA | AB170102.1 | - | 4 - 5 | 534 | 95 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
