Assay Details
Gene Symbol: LMNTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285366.1 | NP_001272295.1 | 5 - 6 | 953 | 63 | |
XM_015430157.1 | XP_015285643.1 | 5 - 6 | 960 | 63 | ||
XM_015430158.1 | XP_015285644.1 | 5 - 6 | 942 | 63 | ||
XM_015430159.1 | XP_015285645.1 | 5 - 6 | 818 | 63 | ||
XM_015430160.1 | XP_015285646.1 | 5 - 6 | 743 | 63 | ||
GenBank mRNA | AB168558.1 | - | 5 - 6 | 953 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
