Assay Details
Gene Symbol: DIMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283491.1 | NP_001270420.1 | 11 - 12 | 1078 | 105 | |
GenBank mRNA | AB060843.1 | - | 11 - 12 | 1078 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
