Assay Details
Gene Symbol: SUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285113.1 | NP_001272042.1 | 3 - 4 | 248 | 65 | |
XM_015451155.1 | XP_015306641.1 | 3 - 4 | 286 | 65 | ||
XM_015451156.1 | XP_015306642.1 | 3 - 4 | 215 | 65 | ||
GenBank mRNA | AB168249.1 | - | 3 - 4 | 248 | 65 | |
AB170806.1 | - | 3 - 4 | 265 | 65 | ||
AB174427.1 | - | 3 - 4 | 248 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
