Assay Details
Gene Symbol: ENSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005541979.2 | XP_005542036.1 | 3 - 4 | 520 | 70 | |
XM_005541980.2 | XP_005542037.1 | 3 - 4 | 524 | 70 | ||
XM_005541982.2 | XP_005542039.1 | 2 - 3 | 331 | 70 | ||
XM_015457710.1 | XP_015313196.1 | 3 - 4 | 421 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
