Assay Details
Gene Symbol: IL22RA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005551951.2 | XP_005552008.1 | 1 - 2 | 205 | 91 | |
XM_005551952.2 | XP_005552009.1 | 1 - 2 | 205 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
