Assay Details
Gene Symbol: NDUFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005555947.1 | XP_005556004.1 | 3 - 4 | 462 | 105 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
