Assay Details
Gene Symbol: NEURL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005558531.2 | XP_005558588.1 | 5 - 6 | 1590 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
