Assay Details
Gene Symbol: GML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005564195.2 | XP_005564252.1 | 3 - 4 | 272 | 85 | |
XM_005564196.2 | XP_005564253.1 | 3 - 4 | 229 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
