Assay Details
Gene Symbol: BIK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005567075.2 | XP_005567132.1 | 2 - 3 | 284 | 114 | |
XM_005567076.2 | XP_005567133.1 | 2 - 3 | 284 | 114 | ||
XM_015457095.1 | XP_015312581.1 | 1 - 2 | 482 | 114 | ||
XM_015457096.1 | XP_015312582.1 | 1 - 2 | 482 | 114 |

More Information
Set Membership: |
![]() ![]() |
