Assay Details
Gene Symbol: ASF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005588233.2 | XP_005588290.1 | 2 - 3 | 1483 | 107 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
