Assay Details
Gene Symbol: BATF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005577339.2 | XP_005577396.1 | 2 - 3 | 875 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005577339.2 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
