Assay Details
Gene Symbol: SLC2A4RG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005569613.2 | XP_005569670.1 | 4 - 5 | 1407 | 83 | |
XM_005569614.1 | XP_005569671.1 | 4 - 5 | 1404 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
