Assay Details
Gene Symbol: ECM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005582250.2 | XP_005582307.1 | 8 - 9 | 1975 | 96 | |
XM_005582251.2 | XP_005582308.1 | 8 - 9 | 1905 | 96 | ||
XM_005582252.2 | XP_005582309.1 | 8 - 9 | 1690 | 96 | ||
XM_015436581.1 | XP_015292067.1 | 9 - 10 | 1771 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
