Assay Details
Gene Symbol: AKAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005593581.2 | XP_005593638.1 | 2 - 3 | 264 | 65 | |
XM_005593582.2 | XP_005593639.1 | 2 - 3 | 267 | 65 | ||
XM_005593583.2 | XP_005593640.1 | 2 - 3 | 241 | 65 | ||
XM_015443650.1 | XP_015299136.1 | 2 - 3 | 241 | 65 | ||
GenBank mRNA | AB168997.1 | - | 2 - 3 | 238 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
