Assay Details
Gene Symbol: BID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005568045.2 | XP_005568102.1 | 4 - 5 | 867 | 86 | |
XM_005568046.2 | XP_005568103.1 | 4 - 5 | 537 | 86 | ||
XM_015457458.1 | XP_015312944.1 | 4 - 5 | 867 | 86 | ||
XM_015457459.1 | XP_015312945.1 | 4 - 5 | 442 | 86 | ||
XM_015457460.1 | XP_015312946.1 | 4 - 5 | 538 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
