Assay Details
Gene Symbol: CTH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005543038.2 | XP_005543095.1 | 3 - 4 | 996 | 100 |
Gene Symbol: LOC102117863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319466.1 | NP_001306395.1 | 466 | 100 | ||
XM_005595676.2 | XP_005595733.2 | 519 | 100 | |||
GenBank mRNA | AB125160.1 | - | 519 | 100 | ||
AY879312.1 | - | 466 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
